Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBMY3AP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_001573.2 | 1 | 47 | |||
GenBank mRNA | U94385.1 | - | LIT2106 RBMIII pseudogene |
1 | 47 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | U94386.3 | - | 1 | 47 | Chr. Y - 9610721 - 9615153 [-] on Build GRCh38 |