Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEPT7P9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027269.1 | 4 | 644 | |||
GenBank mRNA | BC042387.1 | - | 4 | 644 | ||
BC048143.1 | - | 4 | 644 |