Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC39C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243425.1 | NP_001230354.1 | 2 | 1005 | ||
GenBank mRNA | AK074875.1 | - | 2 | 627 |