Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00470 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023925.1 | 1 | 67 | |||
NR_023926.1 | 1 | 67 | ||||
NR_023927.1 | 1 | 67 | ||||
NR_110327.1 | 1 | 67 | ||||
GenBank mRNA | AF295725.1 | - | 1 | 67 | ||
AF295726.1 | - | 1 | 28 | |||
AF295730.1 | - | 1 | 89 |