Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPIEL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003929.2 | 2 | 517 | |||
GenBank mRNA | AK092606.1 | - | 2 | 493 | ||
DA168720.1 | - | 3 | 265 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CN265253.1 | - | 3 | 207 | Chr. 1 - 39529077 - 39559667 [-] on Build GRCh38 |