Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMED10P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002807.4 | 1 | 3447 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | HY307714.1 | - | 1 | 360 | Chr. 8 - 144997968 - 144998455 [+] on Build GRCh38 |