Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015410.1 | 12 | 1781 | |||
GenBank mRNA | AK022865.1 | - | 12 | 1528 | ||
AK026189.1 | - | 12 | 1781 | |||
AK094718.1 | - | 4 | 1029 |