Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC728989 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024442.2 | 4, 5 | 584 | |||
GenBank mRNA | AK299984.1 | - | 4, 5 | 584 |