Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXL20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184906.1 | NP_001171835.1 | 14 | 8163 | ||
NM_032875.2 | NP_116264.2 | 15 | 8259 | |||
XM_005257746.3 | XP_005257803.2 | 15 | 8450 | |||
XM_005257747.3 | XP_005257804.2 | 14 | 8231 |