Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PWARSN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_022011.1 | 1 | 102 | |||
GenBank mRNA | BX644997.1 | - | 1 | 102 |
Gene Symbol: SNRPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AF319522.1 | - | 6 | 842 | ||
AF319524.1 | - | LIT2203 IC-SNURF-SNRPN 3 prime noncoding transcripts |
1 | 88 | ||
U81001.1 | - | LIT2197 IC-SNURF-SNRPN 3 prime noncoding transcripts |
3 | 1519 |