Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCARNA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_004387.1 | 1 | 222 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BQ069333.1 | - | 126 | Chr. - - [-] on Build |