Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002196.2 | 1 | 311 | |||
NR_131223.1 | 1 | 311 | ||||
GenBank mRNA | AL546946.3 | - | 1 | 271 | ||
AL548405.3 | - | 1 | 271 | |||
DA443982.1 | - | 1 | 311 |
Gene Symbol: RPS12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC002550.1 | - | 101 |