Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnm3os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AK051872.1 | - | LIT3445 D230012O04Rik |
1 | 615 |