Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nct1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AY940662.1 | - | LIT3382 Nct1 |
1 | 616 |