Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Malat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | BC039749.1 | - | LIT3351 MALAT-1 |
14 |