Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC287004 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_110637.1 | 1 | 2105 | |||
GenBank mRNA | DQ080211.1 | - | LIT3459 PINC |
1 | 2105 |