Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AY927517.1 | - | LIT3527 AY927517 |
1 | 106 |