Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | XM_011516480.2 | XP_011514782.1 | 2 | 2922 | ||
GenBank mRNA | AY927451.1 | - | LIT3479 AY927451 |
1 | 48 |