Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HERC2P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AF041081.1 | - | LIT2191 HERC2P3 |
31 | 5200 | |
CR749409.1 | - | 24 | 3679 |
Gene Symbol: HERC2P9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036443.1 | 13 | 1862 | |||
GenBank mRNA | AK056876.1 | - | 1 | 1682 | ||
AL832775.1 | - | 1 | 6225 | |||
BC047911.1 | - | 13 | 1862 |