Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFATC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AY927566.1 | - | LIT3567 AY927566 |
1 | 372 |