Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AF400491.1 | - | LIT2211 IC-SNURF-SNRPN 3 prime noncoding transcripts |
1 | 17 |