Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDOAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | M21191.1 | - | LIT2112 ALDOAP2 |
1 | 1826 |