Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPSR1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AY312367.1 | - | LIT3327 AAA1 |
3 | 352 | |
AY321515.1 | - | 3 | 233 | |||
U85653.1 | - | 3 | 315 |