Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256410.1 | NP_001243339.1 | 8 | 4103 | ||
NM_001256411.1 | NP_001243340.1 | 6 | 3949 | |||
NM_001256412.1 | NP_001243341.1 | 5 | 3824 | |||
NM_001256415.1 | NP_001243344.1 | 6 | 3944 | |||
NM_021252.4 | NP_067075.1 | 7 | 4016 | |||
NR_046172.1 | 6 | 3954 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CR610854.1 | - | 1 | 724 | Chr. 10 - 27540527 - 27542214 [+] on Build GRCh38 |