Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017008332.1 | XP_016863821.1 | 7 | 1986 | ||
GenBank mRNA | AK090742.1 | - | 1 | 430 | ||
AK124036.1 | - | 1 | 1002 |