Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00437 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126377.1 | 3 | 865 | |||
GenBank mRNA | BC040923.2 | - | 2 | 639 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM927018.1 | - | 671 | Chr. - - [-] on Build |