Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318137.1 | NP_001305066.1 | 4 | 1384 | ||
NM_017715.3 | NP_060185.2 | 6 | 1636 | |||
GenBank mRNA | AK000223.1 | - | 5 | 1579 | ||
AK123799.1 | - | 1 | 1728 |