Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOP-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB014771.1 | - | 1 | 162 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CU456651.1 | - | 1 | 251 | Chr. 4 - 81468836 - 81469398 [-] on Build GRCh38 |