Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322842.1 | NP_001309771.1 | 2 | 2008 | ||
GenBank mRNA | AK026788.1 | - | 1 | 905 |