Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AF319522.1 | - | 4 | 360 | ||
U81001.1 | - | LIT2197 IC-SNURF-SNRPN 3 prime noncoding transcripts |
1 | 1037 |