Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP2B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011531174.2 | XP_011529476.1 | 22 | 5237 | ||
XM_011531177.2 | XP_011529479.1 | 21 | 5195 | |||
XM_017029553.1 | XP_016885042.1 | 21 | 5140 | |||
GenBank mRNA | AK130194.1 | - | 1 | 853 |