Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUSC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011544651.2 | XP_011542953.1 | 9 | 1871 | ||
XM_011544652.2 | XP_011542954.1 | 10 | 1936 | |||
GenBank mRNA | AK026149.1 | - | 1 | 540 |