Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNNM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005269933.4 | XP_005269990.1 | 3 | 2451 | ||
GenBank mRNA | AK090785.1 | - | 1 | 1355 |