Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSNK1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK096950.1 | - | 1 | 212 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA917526.1 | - | 1 | 212 | Chr. 17 - 82244823 - 82245441 [-] on Build GRCh38 |