Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSNK1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001893.4 | NP_001884.2 | 9 | 3055 | ||
NM_139062.2 | NP_620693.1 | 10 | 3119 | |||
NR_110578.1 | 9 | 2914 | ||||
GenBank mRNA | AB209463.1 | - | 8 | 5455 | ||
AK096950.1 | - | 2 | 2083 | |||
BC047000.1 | - | 1 | 678 |