Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSNK1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001893.4 | NP_001884.2 | 9 | 2008 | ||
NM_139062.2 | NP_620693.1 | 10 | 2072 | |||
NR_110578.1 | 9 | 1867 | ||||
GenBank mRNA | AB091044.1 | - | 9 | 1721 | ||
AB209463.1 | - | 8 | 4408 | |||
AK096950.1 | - | 2 | 1036 | |||
AK128863.1 | - | 3113 | ||||
U29171.1 | - | 9 | 1849 |