Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005267730.3 | XP_005267787.1 | 8 | 2624 | ||
GenBank mRNA | AK057060.1 | - | 1 | 17 | ||
BC080600.1 | - | 9 | 1687 | 4889846 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA032623.1 | - | 1 | 286 | Chr. 14 - 74736490 - 74737067 [+] on Build GRCh38 |