Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGD5-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046251.1 | 2 | 519 | |||
NR_046252.1 | 2 | 509 | ||||
NR_046253.1 | 2 | 486 | ||||
NR_046254.1 | 2 | 482 | ||||
NR_046255.1 | 2 | 245 | ||||
GenBank mRNA | AK055924.1 | - | 2 | 487 | ||
AK096279.1 | - | 2 | 502 | |||
BC008609.1 | - | 2 | 188 | |||
BC039246.1 | - | 2 | 469 | |||
BC056913.1 | - | 2 | 245 | |||
BC092452.1 | - | 2 | 250 |