Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011520276.2 | XP_011518578.1 | 6 | 1892 | ||
GenBank mRNA | AL833465.1 | - | 1 | 2200 |