Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317338.1 | NP_001304267.1 | 8 | 2035 | ||
NM_017898.4 | NP_060368.2 | 8 | 2028 | |||
XM_006711407.2 | XP_006711470.1 | 7 | 2584 | |||
XM_011509684.1 | XP_011507986.1 | 8 | 1552 | |||
XM_017001581.1 | XP_016857070.1 | 5 | 1720 | |||
GenBank mRNA | AK000583.1 | - | 1 | 472 | ||
BC010366.1 | - | 7 | 1363 | |||
BC015829.2 | - | 6 | 1298 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK125512.1 | - | 1 | 3409 | Chr. 1 - 220781213 - 220784803 [+] on Build GRCh38 |