Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFIC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001245002.1 | NP_001231931.1 | 11 | 3546 | ||
NM_001245004.1 | NP_001231933.1 | 10 | 3392 | |||
NM_001245005.1 | NP_001231934.1 | 10 | 3415 | |||
NM_005597.3 | NP_005588.2 | 9 | 3306 | |||
NM_205843.2 | NP_995315.1 | 11 | 3569 | |||
GenBank mRNA | AK129956.1 | - | 1 | 1114 |