Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB27B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004163.4 | NP_004154.2 | 6 | 6348 | ||
XM_005266733.1 | XP_005266790.1 | 7 | 6269 | |||
XM_006722518.1 | XP_006722581.1 | 6 | 2858 | |||
XM_011526131.1 | XP_011524433.1 | 7 | 1435 | |||
XM_017025913.1 | XP_016881402.1 | 9 | 7102 | |||
XM_017025914.1 | XP_016881403.1 | 8 | 6497 | |||
GenBank mRNA | AF131784.1 | - | 1 | 1915 |