Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ICE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC041002.2 | - | 1 | 1271 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM721129.1 | - | 1 | 108 | Chr. 5 - 5488402 - 5489184 [+] on Build GRCh38 |