Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037131.2 | NP_001032208.1 | 18 | 9392 | ||
NM_014914.4 | NP_055729.2 | 17 | 9233 | |||
XM_005246059.4 | XP_005246116.1 | 18 | 8698 | |||
XM_006712234.3 | XP_006712297.1 | 20 | 8933 | |||
XM_006712235.3 | XP_006712298.1 | 19 | 9057 | |||
XM_006712237.3 | XP_006712300.2 | 19 | 9714 | |||
XM_006712239.3 | XP_006712302.2 | 20 | 9729 | |||
XM_011510547.2 | XP_011508849.1 | 19 | 9650 | |||
XM_011510548.2 | XP_011508850.1 | 19 | 9568 | |||
XM_011510549.2 | XP_011508851.1 | 18 | 9531 | |||
XM_017003282.1 | XP_016858771.1 | 19 | 9603 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CR608907.1 | - | 1 | 666 | Chr. 2 - 236129677 - 236131772 [+] on Build GRCh38 |