Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRCH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005269365.2 | XP_005269422.1 | 20 | 8486 | ||
XM_005269367.2 | XP_005269424.1 | 19 | 8414 | |||
XM_006713791.3 | XP_006713854.1 | 21 | 5565 | |||
XM_006713792.3 | XP_006713855.1 | 20 | 5493 | |||
XM_006713793.2 | XP_006713856.1 | 20 | 5457 | |||
XM_017007352.1 | XP_016862841.1 | 18 | 8360 | |||
GenBank mRNA | AK125235.1 | - | 1 | 17 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA559512.1 | - | 1 | 17 | Chr. 3 - 197886457 - 197887271 [+] on Build GRCh38 |