Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD3D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017018543.1 | XP_016874032.1 | 1 | 15 | ||
GenBank mRNA | AK093775.1 | - | 1 | 52 |