Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP5P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK055963.1 | - | 1 | 1590 |
Gene Symbol: LOC105373132 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011544354.2 | XP_011542656.1 | 1 | 2920 |