Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MAGI1 SIRNA Assays ›
Gene Symbol:
MAGI1
Gene Name:
membrane associated guanylate kinase, WW and PDZ domain containing 1
Gene Aliases:
AIP-3, AIP3, BAIAP1, BAP-1, BAP1, MAGI-1, Magi1d, TNRC19, WWP3
Chromosome Location:
Chr.3: 65353525 - 66038836 on Build GRCh38
UniGene ID:
Hs.476636
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
n288418
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MAGI1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
XM_005265563.1
XP_005265620.1
24
7596
XM_005265564.1
XP_005265621.1
24
7593
XM_005265565.1
XP_005265622.1
24
7563
XM_005265566.1
XP_005265623.1
24
7557
XM_005265568.1
XP_005265625.1
23
7554
XM_005265570.1
XP_005265627.1
23
7395
XM_005265571.1
XP_005265628.1
22
7305
XM_005265574.3
XP_005265631.1
25
7682
XM_006713407.1
XP_006713470.1
24
7599
XM_006713408.1
XP_006713471.1
24
7596
XM_006713409.1
XP_006713472.1
24
7596
XM_006713410.1
XP_006713473.1
23
7557
XM_006713411.1
XP_006713474.1
23
7515
XM_006713412.1
XP_006713475.1
23
7509
XM_006713414.3
XP_006713477.1
25
7688
XM_011534236.1
XP_011532538.1
23
7560
XM_011534240.2
XP_011532542.1
12
8955
XM_011534241.2
XP_011532543.1
11
8869
XM_017007497.1
XP_016862986.1
24
7593
XM_017007498.1
XP_016862987.1
24
7590
XM_017007499.1
XP_016862988.1
24
7954
XM_017007500.1
XP_016862989.1
24
7944
XM_017007501.1
XP_016862990.1
23
7476
XM_017007502.1
XP_016862991.1
25
7685
XM_017007503.1
XP_016862992.1
25
7685
XM_017007504.1
XP_016862993.1
25
7682
XM_017007505.1
XP_016862994.1
25
7682
XM_017007506.1
XP_016862995.1
25
7646
XM_017007507.1
XP_016862996.1
22
6637
XM_017007508.1
XP_016862997.1
21
6469
GenBank mRNA
AL050184.1
-
1
629
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: AK147358, AK081387.
Gene Ontology Categories:
Function(s)
Process(es)
protein complex assembly
cell adhesion
cell surface receptor signaling pathway
neuron death
protein binding
ATP binding
protein C-terminus binding
protein complex scaffold
alpha-actinin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0