Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01225 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034112.2 | 8 | 1305 | |||
GenBank mRNA | AK057562.1 | - | 1 | 1463 | ||
BC104454.1 | - | 2 | 644 | |||
BC104455.2 | - | 3 | 383 |