Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PACRGL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011513786.2 | XP_011512088.1 | 11 | 5869 | ||
XM_011513805.2 | XP_011512107.1 | 8 | 5410 | |||
XM_017007747.1 | XP_016863236.1 | 10 | 5624 | |||
XM_017007748.1 | XP_016863237.1 | 10 | 5575 | |||
XM_017007749.1 | XP_016863238.1 | 9 | 5490 | |||
XM_017007751.1 | XP_016863240.1 | 9 | 5692 | |||
XM_017007752.1 | XP_016863241.1 | 8 | 5539 | |||
XM_017007753.1 | XP_016863242.1 | 8 | 5409 | |||
XM_017007754.1 | XP_016863243.1 | 6 | 5466 | |||
XM_017007755.1 | XP_016863244.1 | 6 | 5283 | |||
XM_017007756.1 | XP_016863245.1 | 6 | 5276 | |||
XM_017007757.1 | XP_016863246.1 | 6 | 5212 | |||
XM_017007758.1 | XP_016863247.1 | 5 | 5161 | |||
XM_017007759.1 | XP_016863248.1 | 5 | 5109 |